Learn More
Promega KDR Kinase Enzyme System, 1mg
Catalog No. PRV2682 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV2682 1 mg
1 options
Catalog No. PRV2682 Supplier Promega Supplier No. V2682
Add to Cart
Add to Cart
Request a Quote
Recombinant human KDR (amino acids 789-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. KDR (or kinase insert domain receptor) is a growth factor receptor tyrosine kinase originally isolated from human endothelial cells.
  • Easily Screen and Profile KDR Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C