Learn More
KDR Mouse anti-Human, Clone: OTI12C1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802572

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Vascular endothelial growth factor (VEGF) is a major growth factor for endothelial cells. This gene encodes one of the two receptors of the VEGF. This receptor, known as kinase insert domain receptor, is a type III receptor tyrosine kinase. It functions as the main mediator of VEGF-induced endothelial proliferation, survival, migration, tubular morphogenesis and sprouting. The signalling and trafficking of this receptor are regulated by multiple factors, including Rab GTPase, P2Y purine nucleotide receptor, integrin alphaVbeta3, T-cell protein tyrosine phosphatase, etc.. Mutations of this gene are implicated in infantile capillary hemangiomas.Specifications
KDR | |
Monoclonal | |
Unconjugated | |
KDR | |
CD309, FLK1, VEGFR, VEGFR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
3791 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI12C1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P35968 | |
KDR | |
Human recombit protein fragment corresponding to amino acids 786-1356 of human KDR produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.