Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Keap1 Antibody (OTI1B4), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271496PE
Description
Keap1 Monoclonal specifically detects Keap1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.Specifications
Keap1 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
KEAP1 | |
Full length recombinant protein of human Keap1 (NP_987096) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Monkey | |
IgG1 |
Flow Cytometry | |
OTI1B4 | |
Flow Cytometry | |
INRF2, INrf2MGC9454, kelch-like ECH-associated protein 1, Kelch-like protein 19, KIAA0132MGC4407, KLHL19Cytosolic inhibitor of Nrf2, MGC10630, MGC1114, MGC20887 | |
Mouse | |
Affinity Purified | |
Stem Cell Markers | |
9817.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction