Learn More
Invitrogen™ KEAP1 Monoclonal Antibody (1F10B6)

Description
MA5-17106 targets KEAP1 in indirect ELISA, FACS, ICC, IF and WB applications and shows reactivity with Human and Mouse samples. The MA5-17106 immunogen is purified recombinant fragment of human KEAP1 (amino acids: 380-624) expressed in E. Coli. MA5-17106 detects KEAP1 which has a predicted molecular weight of approximately 69.7kDa.
Specifications
Specifications
| Antigen | KEAP1 |
| Applications | ELISA, Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1F10B6 |
| Concentration | Conc. not determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.03% sodium azide |
| Gene | Keap1 |
| Gene Accession No. | Q14145, Q9Z2X8 |
| Gene Alias | cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.