Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KEAP1 Monoclonal Antibody (7G4B10)
GREENER_CHOICE

Catalog No. PIMA517107
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA517107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA517107 Supplier Invitrogen™ Supplier No. MA517107
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-17107 targets KEAP1 in FACS, IHC, indirect ELISA, and WB applications and shows reactivity with Human samples. The MA5-17107 immunogen is purified recombinant fragment of human KEAP1 (amino acids: 380-624 ) expressed in E. Coli. MA5-17107 detects KEAP1 which has a predicted molecular weight of approximately 69.7kDa.

This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KEAP1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7G4B10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Keap1
Gene Accession No. Q14145
Gene Alias cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein
Gene Symbols Keap1
Host Species Mouse
Immunogen Purified recombinant fragment of human KEAP1 (amino acids: 380-624 ) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9817
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.