Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KEAP1 Monoclonal Antibody (OTI1A1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502056S


Description
KEAP1 Monoclonal antibody specifically detects KEAP1 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
KEAP1 | |
Monoclonal | |
0.75 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13137 | |
Mouse | |
Affinity chromatography | |
RUO | |
10241 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Flow Cytometry, Western Blot | |
OTI1A1 | |
Unconjugated | |
CALCOCO2 | |
cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein | |
Full length human recombinant protein of human CALCOCO2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction