Learn More
KEAP1 Mouse anti-Human, Rat, Clone: 4G10H9, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | KEAP1 |
---|---|
Clone | 4G10H9 |
Concentration | 0.5 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6195
![]() ![]() |
Proteintech Group Inc
600271IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This antibody is a mouse monoclonal antibody raised against residues near the C terminus of human KEAP1.
This gene encodes a protein containing KELCH-1 like domains, as well as a BTB/POZ domain. Kelch-like ECH-associated protein 1 interacts with NF-E2-related factor 2 in a redox-sensitive manner and the dissociation of the proteins in the cytoplasm is followed by transportation of NF-E2-related factor 2 to the nucleus. This interaction results in the expression of the catalytic subunit of gamma-glutamylcysteine synthetase. Two alternatively spliced transcript variants encoding the same isoform have been found for this gene.Specifications
KEAP1 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Cytosolic inhibitor of Nrf2, INrf2, KEAP1, Kelch like protein 19, KIAA0132, KLHL19 | |
KEAP1 | |
IgA | |
Affinity Chromatography | |
Antibody |
4G10H9 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat | |
P57790, Q14145 | |
117519, 9817 | |
KEAP1 Fusion Protein Ag0779 | |
Primary | |
-20°C | |
KEAP1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.