Learn More
Invitrogen™ KEAP1 Recombinant Rabbit Monoclonal Antibody (PSH0-90)

Description
Sequence Similarities: 98% Mouse/Rat. Tissue Specificity: Broadly expressed, with highest levels in skeletal muscle. Positive Control: HepG2 cell lysate, HeLa cell lysate, MCF7 cell lysate, A431 cell lysate, Jurkat cell lysate, HEK-293 cell lysate, Daudi cell lysate, A549 cell lysate, NIH/3T3 cell lysate, PC-12 cell lysate, mouse kidney tissue, rat kidney tissue, PC-12. Subcellular Location: Cytoplasm, Nucleus. Predicted band size: 70 kDa.
Specifications
Specifications
| Antigen | KEAP1 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | PSH0-90 |
| Concentration | 1.07 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | Keap1 |
| Gene Accession No. | P57790, Q14145, Q9Z2X8 |
| Gene Alias | cytosolic inhibitor of Nrf2; INrf2; Keap1; kelch like ECH associated protein 1; Kelch-like ECH-associated protein 1; kelch-like family member 19; kelch-like protein 19; Kiaa0132; KLHL19; mKIAA0132; NRF2 cytosolic inhibitor; ring canal protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.