Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kelch-Like Family Member 21 Antibody (2H4), Novus Biologicals™
SDP

Catalog No. NBP237659 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP237659 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP237659 Supplier Novus Biologicals Supplier No. NBP237659
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kelch-Like Family Member 21 Monoclonal specifically detects Kelch-Like Family Member 21 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Kelch-Like Family Member 21
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2H4
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunohistochemistry-Paraffin 1:200-1:1000
Formulation Ascites with 0.03% Sodium Azide
Gene Alias kelch-like 21 (Drosophila), Kelch-Like Protein 21, KIAA0469, KLHL21
Gene Symbols KLHL21
Host Species Mouse
Immunogen Purified recombinant fragment of human KLHL21 expressed in E. Coli.
Molecular Weight of Antigen 66 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9903
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.