Learn More
Invitrogen™ KELL Monoclonal Antibody (BRIC 203)

Description
MA1-25131 detects KELL from human samples.
Specifications
Specifications
| Antigen | KELL |
| Applications | Flow Cytometry, Hemagglutination Assay, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | BRIC 203 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide |
| Gene | KEL |
| Gene Accession No. | P23276 |
| Gene Alias | CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.