Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KELL Monoclonal Antibody (BRIC203)
GREENER_CHOICE

Catalog No. MA180068
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA180068 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA180068 Supplier Invitrogen™ Supplier No. MA180068
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In haemaggulation tests, MA1-80068 reacts with Kp (a-b+) but not Kp (a+b-). This antibody will agglutinate Kp (a+b-c+) and Kp (a-b-c+) erythrocytes but will not react with Ko erythrocytes. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD238 antibody, clone BRIC203 recognizes human CD238, also known as the kell blood group antigen Kpbc. The kell blood group antigens are carried on the approximately 93 kDa erythrocyte membrane glycoprotein.

This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KELL
Applications Flow Cytometry, Hemagglutination Assay, Immunoprecipitation
Classification Monoclonal
Clone BRIC203
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with ≤100mM glycine and <0.1% sodium azide
Gene KEL
Gene Accession No. P23276
Gene Alias CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470
Gene Symbols KEL
Host Species Mouse
Immunogen Human erythrocytes.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3792
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.