Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KELL Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119242
This item is not returnable.
View return policy
Description
KELL Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases.
Specifications
KELL | |
Polyclonal | |
Unconjugated | |
KEL | |
CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
23925 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
Q9EQF2 | |
KEL | |
Recombinant protein Kell Protein. The antigen corresponds to amino acid range 49-358 of the target protein. | |
200 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction