Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KELL Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587644
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587644 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587644 Supplier Invitrogen™ Supplier No. PA587644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QNCGPRPCET SVCLDLRDHY LASGNTSVAP CTDFFSFACG RAKETNNSFQ ELATKNKNRL RRILEVQNSW HPGSGEEKAF QFYNSCMDTL AIEAAGTGPL RQVIEELGGW RISGKWTSLN FNRTLRLLMS QYGHFPFFRA YLGPHPASPH TPVIQIDQPE FDVPLKQDQE QKIYAQIFRE YLTYLNQLGT LLGGDPSKVQ EHSSLSISIT SRLFQFLRPL EQRRAQGKLF QMVTIDQLKE M; Positive Samples: Raji, A-375, K-562; Cellular Location: Cell membrane, Single-pass type II membrane protein.

This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KELL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.84 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KEL
Gene Accession No. P23276
Gene Alias CD238; ECE3; Kel; Kel blood group protein; Kell blood group; kell blood group antigen; kell blood group glycoprotein; Kell blood group glycoprotein homolog; Kell blood group, metallo-endopeptidase; Kell protein; RGD1565470
Gene Symbols KEL
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 70-310 of human KEL (NP_0004111).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3792
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.