Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kemtec™ AP™ Chemistry Kits: Determining Molar Enthalpy Using Hess's Law
SCI_ED

Catalog No. S07339 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Grade:
10 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S07339 10 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S07339 Supplier Aqua Phoenix Scientific Supplier No. 16112
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Three bottles; Hydrochloric Acid, 1.0N; Magnesium Ribbon, 2g; Magnesium Oxide, 5g

Requires

Thermometers, balances, basic lab supplies and styrofoam cups

This calorimetry lab uses reliable reactions as students measure temperature changes, determine the calorimeter constant and ultimately calculate the molar enthalpy of formation for magnesium oxide using Hess's Law.

Students compare lab results to standard values.

  • Ideal for Honors or AP Chemistry
  • Instructor's manual features background material, easy to follow stepwise protocols, assessment questions, sample data, modeled calculations, and an answer key
  • Procedures can easily be divided or used as collaborative lessons
  • Kit designed to exceed 2013 AP Next Generation National Standards
  • Meets AP Science Practices 2 & 7
  • Targets Grades: 10-14
  • Accommodates 24 Students Working in Groups of two

Kemtec AP kits are compatible with optional data collection and simulations using TI-Nspire handhelds and software. Free activities can be found at www.fisheredu.com/TI.

Specifications

Certifications/Compliance Meets new 2013 AP standards, meets AP science practices 2 and 7
Grade 10 to 14
For Use With (Application) For students measure temperature changes, determine the calorimeter constant and ultimately calculate the molar enthalpy of formation for magnesium oxide using hess's law
Includes Three bottles, 1n hydrochloric acid, 2g magnesium ribbon, 5g magnesium oxide
Class Size 24 Students (12 Groups)
Product Type Determining Molar Enthalpy Using Hess'S Law Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.