Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kemtec™ Matters for Minors Kit

Click to view available options
Class Size:
24 Students
Description
Includes
Instructor manual, sodium bicarbonate, white vinegar, isopropyl alcohol, plastic cups, plastic funnels, shells, colored blocks, balloons, sets of plastic containers of different shapes, rocks, paper objects, metal and plastic objects, plastic bags, balls, marbles, candles, wood splints, pill vials, pieces of cloth, plastic circles, buoyant items, rubber bands, cotton balls
Requires
Hot plate, assorted containers and ice
- Explore three states of matter (solids, liquids and gases) and associated properties
- Extensive materials included for sorting and classifying solids
- Chemicals included for exploring gas phases of matter and changes in matter
- The kit stresses a discovery approach to the topics in the exercise for both the students and the instructor
- Grades K to 4
- Class size: 24 students
Specifications
Specifications
Product Type | Matters For Minors Kit |
Grade | 2 to 3 |
For Use With (Application) | An introductory study of chemistry |
Includes | Instructor manual, sodium bicarbonate, white vinegar, isopropyl Alcohol, Plastic cups, Plastic funnels, shells, colored blocks, balloons, sets of Plastic containers of different shapes, rocks, paper objects, Metal and Plastic objects, Plastic bags, balls, marbles, candles, Wood splints, pill vials, pieces of cloth, Plastic circles, buoyant items, Rubber bands, Cotton balls |
Class Size | 24 Students |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction