Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Keratin 31 Antibody (LHTric-1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $499.50
Specifications
Antigen | Keratin 31 |
---|---|
Clone | LHTric-1 |
Concentration | 1.0 mg/mL |
Applications | Western Blot, Immunohistochemistry |
Classification | Monoclonal |
Description
Keratin 31 Monoclonal specifically detects Keratin 31 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Keratin 31 | |
1.0 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Rat | |
Hair Keratin, Type I Ha, Hard Keratin, Type I, 1, hHa1, K31, Keratin 31, Keratin, Hair, Acidic, 1, Keratin, Type I Cuticular Ha1, Keratin-31, KRTHA1 | |
KRT31 | |
IgG2a | |
Protein A purified |
LHTric-1 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Cancer, Cell Cycle and Replication | |
PBS with 0.02% Sodium Azide | |
3881 | |
CVPRPRCGPCNSFVR | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title