Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Keratin 31 Antibody (LHTric-1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253114IR
Description
Keratin 31 Monoclonal specifically detects Keratin 31 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Keratin 31 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
KRT31 | |
CVPRPRCGPCNSFVR | |
0.1 mL | |
Cancer, Cell Cycle and Replication | |
3881 | |
Store at 4C in the dark. | |
IgG2a |
Western Blot, Immunohistochemistry | |
LHTric-1 | |
Western Blot, Immunohistochemistry | |
Hair Keratin, Type I Ha, Hard Keratin, Type I, 1, hHa1, K31, Keratin 31, Keratin, Hair, Acidic, 1, Keratin, Type I Cuticular Ha1, Keratin-31, KRTHA1 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction