Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Keratin 31 Antibody (LHTric-1), mFluor Violet 450 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP253114MFV450
Description
Keratin 31 Monoclonal antibody specifically detects Keratin 31 in Human, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| Keratin 31 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified | 
| Western Blot, Immunohistochemistry | |
| LHTric-1 | |
| Western Blot, Immunohistochemistry | |
| Hair Keratin, Type I Ha, Hard Keratin, Type I, 1, hHa1, K31, Keratin 31, Keratin, Hair, Acidic, 1, Keratin, Type I Cuticular Ha1, Keratin-31, KRTHA1 | |
| CVPRPRCGPCNSFVR | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 3881 | |
| Store at 4C in the dark. | |
| IgG2a | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            