Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Keratin 32 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568042
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568042 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568042 Supplier Invitrogen™ Supplier No. PA568042
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keratin 32 Polyclonal Antibody for Western Blot

The protein encoded by this gene is a member of the keratin gene family. As a type I hair keratin, it is an acidic protein which heterodimerizes with type II keratins to form hair and nails. The type I hair keratins are clustered in a region of chromosome 17q12-q21 and have the same direction of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Keratin 32
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KRT32
Gene Accession No. Q14532
Gene Alias HA2; hair keratin; Hair keratin, type I Ha2; hard keratin, type I, 2; HHA2; HKA2; K32; Ka26; keratin 32; keratin 32, type I; keratin, hair, acidic, 2; keratin, type I cuticular Ha2; Keratin-32; KRT32; KRTHA2; type I cuticular hair keratin; type I hair keratin KA26
Gene Symbols KRT32
Host Species Rabbit
Immunogen Synthesized peptide derived from human Keratin 32.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3882
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.