Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Keratin 36 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | Keratin 36 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
Keratin 36 Polyclonal specifically detects Keratin 36 in Human samples. It is validated for Western Blot.Specifications
Keratin 36 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
8689 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
HA6, Hair keratin, type I Ha6, HHA6, HKA6, K36, keratin 36, keratin, hair, acidic, 6, keratin-36, KRTHA6keratin, type I cuticular Ha6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 388-467 of human Keratin 36 (NP_003762.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title