Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin 39 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9826 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9826 Supplier Bioss Supplier No. BS16950R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRT39 is a protein coding gene.

Specifications

Antigen Keratin 39
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene KRT39
Gene Accession No. Q6A163
Gene Alias 4732494G06Rik; CK-39; Cytokeratin-39; K39; Ka35; keratin 39; keratin 39, type I; Keratin, type I cytoskeletal 39; Keratin-39; KRT39; LOC100340693; ortholog of type I hair keratin KA35; Type I hair keratin Ka35; type I hair keratin KA35-like
Gene Symbols KRT39
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Keratin 39.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 390792
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.