Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin 80 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989827
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9827 Supplier Bioss Supplier No. BS16952R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into epithelial keratins and hair keratins. This gene′s expression profile shows that it encodes a type II epithelial keratin, although structurally the encoded protein is more like a type II hair keratin. This protein is involved in cell differentiation, localizing near desmosomal plaques in earlier stages of differentiation but then dispersing throughout the cytoplasm in terminally differentiating cells. The type II keratins are clustered in a region of chromosome 12q13. Two transcript variants encoding two different fully functional isoforms have been found for this gene.

Specifications

Antigen Keratin 80
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene KRT80
Gene Accession No. Q6KB66
Gene Alias 1200016G03Rik; 2310041I20Rik; CK-80; Cytokeratin-80; K80; Kb2; Kb20; keratin 80; keratin 80, type II; keratin b20; Keratin, type II cytoskeletal 80; Keratin-80; KRT80; type II keratin; type II keratin Kb20; type II keratin-20; Type-II keratin Kb20
Gene Symbols KRT80
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Keratin 80.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 144501
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.