Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keratin82 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501987619
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-7619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-7619 Supplier Bioss Supplier No. BS6947R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KRT82 is a member of the keratin gene family. As a type II hair keratin, it is a basic protein which heterodimerizes with type I keratins to form hair and nails. The type II hair keratins are clustered in a region of chromosome 12q13 and are grouped into two distinct subfamilies based on structure similarity. One subfamily, consisting of KRTHB1, KRTHB3, and KRTHB6, is highly related. The other less-related subfamily includes KRTHB2, KRTHB4, and KRTHB5. All hair keratins are expressed in the hair follicle; this keratin appears to be a hair cuticle-specific keratin.

Specifications

Antigen Keratin82
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene KRT82
Gene Accession No. Q9NSB4
Gene Alias hard keratin, type II, 2; HB2; Hb-2; K82; keratin 82; keratin 82, type II; keratin complex 2, basic, gene 20; keratin, hair, basic, 2; Keratin, type II cuticular Hb2; Keratin-82; KRT K82; Krt2-20; KRT82; KRTHB2; Type II hair keratin Hb2; Type-II keratin Kb22
Gene Symbols KRT82
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Keratin82/KRT82.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3888
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.