Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kerosene, Odorless, Thermo Scientific Chemicals

Catalog No. p-152606
Click to view available options
Quantity:
1 L
4 L

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8008-20-6
MDL Number MFCD00135561

Specifications

Density 4.5 (Air = 1.0)
Quantity 1 L
Merck Index 15, 5342
Vapor Pressure 5mmHg at 38°C
Chemical Name or Material Kerosene
DOTInformation DOT Class 3
EINECSNumber 232-366-4
RTECSNumber OA5500000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.