Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketamine Mouse anti-Human, Clone: 4E2, Bioss
SDP

Catalog No. 501979905
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-197-9905 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-197-9905 Supplier Bioss Supplier No. BSM2069M
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ketamine is a drug used in human and veterinary medicine. Its hydrochloride salt is sold as Ketanest, Ketaset, and Ketalar. Pharmacologically, ketamine is classified as an NMDA receptor antagonist.[2] At high, fully anesthetic level doses, ketamine has also been found to bind to opioid Mu receptors and sigma receptors.[3][4] Like other drugs of this class such as tiletamine and phencyclidine (PCP), it induces a state referred to as 'dissociative anesthesia'[5] and is used as a recreational drug.

Specifications

Antigen Ketamine
Applications ELISA
Classification Monoclonal
Clone 4E2
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Host Species Mouse
Immunogen KLH conjugated Ketamine.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.