Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Ketanserin Monoclonal Antibody (KET101)
GREENER_CHOICE

Catalog No. 01671810
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-810 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-810 Supplier Invitrogen™ Supplier No. MA110780
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10780 detects Ketanserin. MA1-10780 has been successfully used in ELISA applications. The MA1-10780 immunogen is Ketanserin, conjugated to KLH.

Ketanserin is a quinazoline derivative and serotonin (5-hydroxytryptamine, 5HT) receptor subtype 2 (5-HTR2) antagonist with potential antihypertensive and antiplatelet activities. Following administration, ketanserin binds to and inhibits the signaling mediated by 5-HTR2, which inhibits serotonin-dependent vasoconstriction and platelet activation. [National Center for Biotechnology Information. PubChem Compound Database; CID=3822].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ketanserin
Applications ELISA
Classification Monoclonal
Clone KET101
Concentration ∼1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Alias C22H22FN3O3; Ketanserin tartrate; Ketanserinum; Ketaserin
Host Species Mouse
Immunogen Ketanserin, conjugated to KLH.
Purification Method Purified
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.