Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Ketanserin tartrate
GREENER_CHOICESDP

Catalog No. 090850 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-085-0 50 mg
09-082-50 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 09-085-0 Supplier R&D Systems™ Supplier No. 0908/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective 5-HT2A antagonist. Also antagonist at 5-HT1D

Selective 5-HT2A serotonin receptor antagonist; can also be used to discriminate between 5-HT1D and 5-HT1B receptor subtypes.

Chemical Identifiers

CAS 83846-83-7
Molecular Formula C26H30FN3O10

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 545.52
Purity >97%
Molecular Weight (g/mol) 563.54
Chemical Name or Material Ketanserin tartrate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.