Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoconazole, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18603640 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Poly Pail

K1149, 65277-42-1, C26H28Cl2N4O4

Spectrum™ Chemical Ketoconazole, USP is a synthetic antifungal medication that is used primarily to treat fungal infections. While its use as an oral tablet has been discontinued in many countries due to concerns that it caused serious liver injury, it is still used in a variety of topical administrations such as creams, which are used to treat tinea, candidal paronychia, and pityriasis versicolor and in shampoos used primarily to treat seborrheic dermatitis (dandruff). The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 65277-42-1
Molecular Formula C26H28Cl2N4O4
Molecular Weight (g/mol) 531.43
InChI Key XMAYWYJOQHXEEK-ZEQKJWHPSA-N
IUPAC Name 1-[4-(4-{[(2S,4R)-2-(2,4-dichlorophenyl)-2-[(1H-imidazol-1-yl)methyl]-1,3-dioxolan-4-yl]methoxy}phenyl)piperazin-1-yl]ethan-1-one
SMILES CC(=O)N1CCN(CC1)C1=CC=C(OC[C@@H]2CO[C@](CN3C=CN=C3)(O2)C2=CC=C(Cl)C=C2Cl)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 1 kg
UN Number UN2811
Specific Rotation -1° to +1°
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.