Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
α-Ketoglutaric Acid Analytical Standard, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mg
50 mg
Specifications
Specifications
CAS | 328-50-7 |
Flash Point | Not applicable |
Molecular Formula | C5H6O5 |
Linear Formula | HOOCCH2CH2COCOOH |
MDL Number | MFCD00004165 |
Quantity | 10 mg |
UN Number | NONH for all modes of transport |
Synonym | 2-Oxoglutaric Acid; 2-Oxopentanedioic Acid |
Formula Weight | 146.10 |
Percent Purity | 99% to 101% (T) |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction