Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ketohexokinase Antibody (OTI1D8), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271069MFV500
Description
Ketohexokinase Monoclonal antibody specifically detects Ketohexokinase in Human samples. It is validated for Flow CytometrySpecifications
Ketohexokinase | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
OTI1D8 | |
Flow Cytometry | |
EC 2.7.1.3, Hepatic fructokinase, ketohexokinase, ketohexokinase (fructokinase) | |
Full length human recombinant protein of human KHK (NP_000212) produced in HEK293T cell. | |
0.1 mL | |
Lipid and Metabolism | |
3795 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction