Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketoprofen, Micronized, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18603647
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Poly Bottle

K1392, 22071-15-4, C16H14O3

Spectrum™ Chemical Ketoprofen, Micronized, USP

Chemical Identifiers

CAS 22071-15-4
Molecular Formula C16H14O3
Molecular Weight (g/mol) 254.29
InChI Key DKYWVDODHFEZIM-UHFFFAOYNA-N
IUPAC Name 2-(3-benzoylphenyl)propanoic acid
SMILES CC(C(O)=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Melting Range 92.0-97.0°C
Packaging Poly Bottle
Quantity 1 kg
UN Number UN2811
Specific Rotation +1° to -1°
Percent Purity 98.5 to 101%
Grade USP
Physical Form Micronized Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.