Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Ketoprofen Polyclonal Antibody
Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA175229
Description
Working Concentration: 1.25 μg/mL. Sensitivity: 10 ng/mL Ketoprofen produces 28% inhibition in a competitive ELISA employing Ketoprofen polyclonal antibody.
Ketoprofen is only found in individuals that have used or taken this drug. It is a propionic acid derivative and is a nonsteroidal anti-inflammatory agent (NSAIA) with analgesic and antipyretic properties. The anti-inflammatory effects of ketoprofen are believed to be due to inhibition cylooxygenase-2 (COX-2), an enzyme involved in prostaglandin synthesis via the arachidonic acid pathway. This results in decreased levels of prostaglandins that mediate pain, fever and inflammation. Ketoprofen is a non-specific cyclooxygenase inhibitor and inhibition of COX-1 is thought to confer some of its side effects, such as GI upset and ulceration. Ketoprofen is thought to have anti-bradykinin activity, as well as lysosomal membrane-stabilizing action. Antipyretic effects may be due to action on the hypothalamus, resulting in an increased peripheral blood flow, vasodilation, and subsequent heat dissipation. [National Center for Biotechnology Information. PubChem Compound Database; CID=3825].
Specifications
Ketoprofen | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.3 | |
Sheep | |
purified | |
RUO | |
Chemical | |
Antibody | |
IgG |
ELISA | |
Unconjugated | |
C16H14O3 | |
Ketoprofen-BTG. | |
1 mg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction