Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ketotifen Hydrogen Fumarate, EP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18603668 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-668 5 g Amber Glass Bottle
18-603-664 1 g Amber Glass Bottle
18-603-665 25 g Amber Glass Bottle
18-603-666 100 g Amber Glass Bottle
18-603-667 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-603-668 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. K30125GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

K3012, 34580-14-8, C19H19NOS·C4H4O4

Spectrum™ Chemical Ketotifen Hydrogen Fumarate, EP is used to treat allergic conjunctivitis and to prevent asthma attacks. The EP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 34580-14-8
Molecular Formula C23H23NO5S
Molecular Weight (g/mol) 425.50
InChI Key YNQQEYBLVYAWNX-WLHGVMLRSA-N
IUPAC Name (2E)-but-2-enedioic acid; 2-(1-methylpiperidin-4-ylidene)-6-thiatricyclo[8.4.0.03,7]tetradeca-1(14),3(7),4,10,12-pentaen-8-one
SMILES OC(=O)\C=C\C(O)=O.CN1CCC(CC1)=C1C2=C(SC=C2)C(=O)CC2=CC=CC=C12

Specifications

CAS Max % 100%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98.5 to 101%
Grade EP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.