accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34580-14-8

IUPAC Name: (2E)-but-2-enedioic acid; 2-(1-methylpiperidin-4-ylidene)-6-thiatricyclo[8.4.0.0³,⁷]tetradeca-1(14),3(7),4,10,12-pentaen-8-one
Synonyms: ketotifen
Molecular Weight (g/mol): 425.5
Molecular Formula: C23H23NO5S
InChi Key: YNQQEYBLVYAWNX-WLHGVMLRSA-N
SMILES: OC(=O)\C=C\C(O)=O.CN1CCC(CC1)=C1C2=C(SC=C2)C(=O)CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (1)
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (5)
  • (2)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (5)
  • (2)
  • (2)
  • (1)
  • (5)
  • (5)
  • (5)
  • (2)
  • (1)
  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (5)
  • (2)
  • (5)
  • (1)