Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alconox Inc. Keylajet™ Low-Foaming High Alkaline Liquid Detergent
GREENER_CHOICE

Catalog No. 18111008 Shop All Alconox Products
Click to view available options
Volume (English):
5 gal.
15 gal.
55 gal.
265 gal.
This item is not returnable. View return policy
This item is not returnable. View return policy

For Tough-to-Clean Residues on Hard Surfaces

  • Low-Foaming Chelating Alkaline Detergent
  • Concentrated, corrosion-inhibited formula
  • Caustic liquid detergent with chelators
  • Free rinsing to provide reliable results and no interfering residues
  • For hard to clean soils, grit, grime, slime, grease, buffing compounds, oils, particulates, deposits, chemical and solvents, mold releases, metal oxides, and much more

Specifications

Product Type Detergent
pH 12
Sterility Non-sterile
Appearance Pale Yellow
Container Type Drum
Content And Storage Closed, Upright in a Cool, Dry Place (15°C to 30°C)
Description Low-Foaming, High Alkaline Liquid
Filtered Filtered
For Use With (Application) Removing Hard to Clean Soils, Grit, Grime, Slime, Grease, Buffing Compounds, Oils, Particulates, Deposits, Chemical/Solvents, Mold Releases, Metal Oxides, and Much More
For Use With (Equipment) Manufacturing Equipment, Cosmetics Residues, Injection Molds, Fouled Metal Parts in Marina, Automotive, Aerospace, and Oil/Gas Maintenance. Repair Operations Such as Filters, Fins, Exchangers, Pump Heads, Steel Engine Blocks, Exhaust Systems, and More. Metal Castings, Forgings/Stampings, Industrial Parts, Pipes, Tanks and Reactors. Use to Clean Steel Prior to Passivation.
Quantity 1 Ea.
Volume (English) 55 gal.
Volume (Metric) 208 L
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.