Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Keystone Safety Nylon Beard Nets, 1/8 in. Holes

Click to view available options
:
White; Large
Description
- Beard net
- Soft heavyweight nylon, 1/8 in. hole
- Honeycomb style
- Flame-resistant
- Large , white
Specifications
Specifications
| Type | General Purpose Beard Cover |
| Size | Large |
| Material | Nylon |
| Color | White |
| Disposable | Single-use |
| For Use With (Application) | Arc Flash Industry, Electrical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction