Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Nylon Beard Nets, 1/16 in. Holes
SDP

Catalog No. p-4014918
Click to view available options
Color:
Blue
White
Size:
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Soft honeycomb nylon beard nets provide flame-resistant protection.

  • Beard net
  • Soft heavyweight nylon, 1/16 in. hole
  • Honeycomb style
  • Flame-resistant
  • Available in large, x-large sizes
  • Available in white or blue color

Specifications

Type General Purpose Beard Cover
Size Large
Material Nylon
Color Blue
Disposable Single-use
For Use With (Application) Arc Flash Industry, Electrical
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.