Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Super Sticky™ Polypropylene Shoe Cover
SDP

Catalog No. p-4706506
Click to view available options
Color:
Blue
White
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Water-resistant cover with non-skid bottom.

  • Available in different sizes and colors
  • Made up of polypropylene

Specifications

Size Medium
Material Polypropylene
Coating EVA-PE Film
Color White
Sole Type Non-skid
Construction Polypropylene
For Use With (Application) Manufacturing, food service, industrial, medical, clean room, pharmaceutical
Product Type Controlled Environments Shoe Covers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.