Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chromium Trioxide (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. A100212 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1333-82-0
Molecular Formula CrO3
Molecular Weight (g/mol) 99.99
MDL Number MFCD00010952
InChI Key WGLPBDUCMAPZCE-UHFFFAOYSA-N
Synonym chromium trioxide, chromium vi oxide, monochromium trioxide, chromtrioxid, chromic trioxide, chromsaeureanhydrid, anhydride chromique, chromium 6+ oxide, chromium vi trioxide, chromium oxide
PubChem CID 14915
ChEBI CHEBI:48240
IUPAC Name trioxochromium
SMILES O=[Cr](=O)=O

Specifications

Melting Point 196°C
Color Violet
pH 1
Physical Form Solid
Quantity 2.5 kg
Assay Percent Range ≥98.0 %
Formula Weight 99.99
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Density 2.700g/cm³
Chemical Name or Material Chromium Trioxide
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Explosive when mixed with combustible material. May be fatal if inhaled. Causes severe burns by all exposure routes. Toxic in contact with skin and if swallowed. May cause sensitization by inhalation and skin contact. Danger of serious damage to health by prolonged exposure. May cause heritable genetic damage. Possible risk of impaired fertility. Hygroscopic. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Wear personal protective equipment. Use only under a chemical fume hood. Use only under a chemical fume hood. Keep away from clothing and other combustible materials.

NFPA

Health:3
Flammability:0
Instability:1
DOTInformation DOT Class 5.1, : Oxidizer
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.