Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
Citric Acid Monohydrate (Granular/USP), Fisher Chemical™
Shop All Fisherbrand Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                3 kg
                            
                        
                            
                                10 kg
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Poly Bottle
                            
                        
                            
                                Poly Pail
                            
                        
                    Specifications
Specifications
| pH | 2.2 | 
| Melting Point | 135°C to 152°C | 
| Boiling Point | 175°C | 
| Color | White | 
| Formula Weight | 210.14 | 
| Percent Purity | 99.5 to 100.5% (USP) | 
| Grade | USP | 
| Density | 1.54g/mL | 
| Quantity | 3 kg | 
| Flash Point | 174°C | 
| Show More | 
Safety and Handling
 
                        
                        | Hazard Statement | WARNING! | 
| Precautionary Statement | Emergency Overview Risk of serious damage to eyes. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. Obtain medical attention. . NFPA Health:2 Flammability:1 Instability:0 | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                 
                                