Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Oleic Acid (NF/FCC), Fisher Chemical
Shop All Fisherbrand Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
| Acidity | 196 to 204 (FCC/NF) |
| Melting Point | 13°C |
| Color | Colorless |
| Density | 0.890g/cm³ |
| Boiling Point | 360°C |
| Ignition Residue | 0.01% max. (FCC/NF) |
| Packaging | Glass Bottle |
| Linear Formula | CH3(CH2)7CH:CH(CH2)7CO2H |
| Quantity | 500 mL |
| Saponification Value | 196 to 206 (FCC) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction