Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
1-Butanol (HPLC), Fisher Chemical™
Shop All Fisherbrand Products
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                1 L
                            
                        
                            
                                4 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                EcoSafPak™ Amber Glass Bottle
                            
                        
                            
                                Safe-Cote™ Amber Glass 
                            
                        
                            
                                Wide Neck Amber Bottle
                            
                        
                    Specifications
Specifications
| Melting Point | -89°C | 
| Absorbance | 1.00 AU max. at 215nm, .50 AU max. at 220nm, 0.20 AU max. at 230nm, 0.025 AU max. at 254nm | 
| Color | Colorless | 
| Density | 0.810g/cm³ | 
| Boiling Point | 117.6°C | 
| Quantity | 1 L | 
| Identification | Pass Test | 
| Packaging | Safe-Cote™ Amber Glass | 
| Physical Form | Liquid | 
| Refractive Index | 1.3952 to 1.3992 (at 25°C) | 
| Show More | 
Safety and Handling
 
                        
                        
                             
                        
                        | Hazard Statement | WARNING! | 
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. Risk of serious damage to eyes. Irritating to eyes, respiratory system and skin. Vapors may cause drowsiness and dizziness. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:2 Flammability:3 Instability:0 | 
| DOTInformation | DOT Class 3, : Flammable Liquid | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                 
                                