Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, Anhydrous (Histological), Fisher Chemical™
GSA_VA

Catalog No. A40520 Shop All Fisherbrand Products
Click to view available options
Quantity:
1 gal.
4 L
20 L
Packaging:
Lined Steel Drum
Poly Bottle

Used in histology, pathology, and cell biology applications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5, 108-10-1, 141-78-6, 67-56-1, 108-88-3
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -90°C
Density 1.5 g/mol (Vapor Density)
Boiling Point 77.1°C
Quantity 20 L
Identification Pass Test
Packaging Lined Steel Drum
Physical Form Liquid
Specific Gravity 0.785 to 0.792 (at 25°C)
Vapor Pressure 48mmHg
UN Number 1170
Water 0.5% max.
Grade Histological
Concentration or Composition (by Analyte or Components) Ethyl Alcohol, 95.27%; Methyl Isobutyl Ketone, 1.0%; Ethyl Acetate, 1.0%; Hydrocarbon, 1.0%
Chemical Name or Material Ethanol, Anhydrous
Show More Show Less
DOTInformation DOT Class 3, : Flammable Liquid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.