Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, Anhydrous (Histological), Fisher Chemical™
GSA_VA

Catalog No. A405P4 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
4 L
20 L
Packaging:
Lined Steel Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A405P-4 4 L Poly Bottle
A405-20 20 L Lined Steel Drum
A405F-1GAL 1 gal. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A405P-4
Only null left
Add to Cart

Used in histology, pathology, and cell biology applications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 108-10-1 , 141-78-6 , 67-56-1 , 108-88-3
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -90°C
Quantity 4 L
Identification Pass Test
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.785 to 0.792 (at 25°C)
Vapor Pressure 48mmHg
UN Number 1170
Water 0.5% max.
Boiling Point 77.1°C
Grade Histological
Concentration or Composition (by Analyte or Components) Ethyl Alcohol, 95.27%; Methyl Isobutyl Ketone, 1.0%; Ethyl Acetate, 1.0%; Hydrocarbon, 1.0%
Chemical Name or Material Ethanol, Anhydrous
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 3, : Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.