Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Propanol (Spectranalyzed™), Fisher Chemical™
Shop All Fisherbrand Products
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Specifications
Specifications
| pH | 7 |
| Melting Point | -89.5°C |
| Absorbance | Pass Test |
| Color | Colorless |
| Density | 0.785g/cm³ |
| Boiling Point | 81°C |
| Quantity | 4 L |
| Identification | Pass Test |
| Infrared Spectrum | Conforms to reference |
| Packaging | Glass Bottle |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Aspiration hazard if swallowed - can enter lungs and cause damage. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen. NFPA Health:1 Flammability:3 Instability:0 |
| DOTInformation | DOT Class 3, : Flammable Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction