Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ammonium Phosphate Monobasic (Crystalline/Certified ACS), Fisher Chemical™
GSA_VA

Catalog No. A6843 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
Poly Bottle

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Melting Point 190°C
Color White
pH 3.8 to 4.4
Physical Form Solid
Quantity 3 kg
Assay Percent Range ≥98.0%
Linear Formula NH4H2PO4
Formula Weight 115.03
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Decomposition Information >190°C
Density 1.8g/mL
Chemical Name or Material Ammonium Phosphate Monobasic
Show More Show Less
Hazard Statement CAUTION!
Precautionary Statement Emergency Overview
May cause skin, eye, and respiratory tract irritation. The toxicological properties have not been fully investigated. Wear personal protective equipment. Ensure adequate ventilation. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. Use personal protective equipment. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. Move to fresh air. If breathing is difficult, give oxygen.

NFPA

Health:1
Flammability:0
Instability:0
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.