Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Citric Acid, Anhydrous (Crystalline/USP), Fisher Chemical™

Catalog No. A95250LB Shop All Fisherbrand Products
Click to view available options
Quantity:
3 kg
250 lb.
Packaging:
Fiber Drum
Poly Bottle

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

pH 1.7
Melting Point 153°C
Boiling Point 175°C
Color White
Formula Weight 192.13
Percent Purity 99.5 to 100.5% (USP)
Grade USP
Density 1.67g/mL
Quantity 250 lb.
Ignition Residue 0.1% max. (USP)
Packaging Fiber Drum
Physical Form Solid
Chemical Name or Material Citric Acid, Anhydrous
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.