Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Acetylpyridine, 98%

Catalog No. AC102685000
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

CAS: 1122-62-9 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-62-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
InChI Key AJKVQEKCUACUMD-UHFFFAOYSA-N
Synonym 2-acetylpyridine, 1-pyridin-2-yl ethanone, methyl 2-pyridyl ketone, 2-acetopyridine, 2-pyridyl methyl ketone, acetylpyridine, ethanone, 1-2-pyridinyl, ketone, methyl 2-pyridyl, 1-pyridin-2-yl ethan-1-one, 1-2-pyridinyl ethanone
PubChem CID 14286
IUPAC Name 1-pyridin-2-ylethanone
SMILES CC(=O)C1=CC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 8.0°C to 10.0°C
Color Colorless to Brown
Density 1.0800g/mL
Boiling Point 188.0°C to 189.0°C
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5200 to 1.5220
Linear Formula C5H4NCOCH3
Quantity 500 g
Specific Gravity 1.08
Solubility Information Solubility in water: 200g/L (20°C). Other solubilities: miscible with ethanol
Formula Weight 121.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Acetylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 214-355-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.