Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylpyridine, 98%

Catalog No. AC102701000
Click to view available options
Quantity:
10 g
100 g
500 g
Packaging:
Glass bottle

CAS: 1122-54-9 | C7H7NO | 121.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-54-9
Molecular Formula C7H7NO
Molecular Weight (g/mol) 121.14
MDL Number MFCD00006433
InChI Key WMQUKDQWMMOHSA-UHFFFAOYSA-N
Synonym 4-acetylpyridine, methyl 4-pyridyl ketone, ethanone, 1-4-pyridinyl, ketone, methyl 4-pyridyl, 1-pyridin-4-yl ethan-1-one, 1-pyridin-4-yl ethanone, pyridine, 4-acetyl, 4-acetyl pyridine, 4-pyridyl methyl ketone, unii-g47437iow7
PubChem CID 14282
IUPAC Name 1-pyridin-4-ylethanone
SMILES CC(=O)C1=CC=NC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 13.0°C to 16.0°C
Color Brown to Yellow
Density 1.0900g/mL
Boiling Point 212.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5270 to 1.5310
Linear Formula (C5H4N)COCH3
Quantity 100 g
Beilstein 21, 279
Specific Gravity 1.09
Solubility Information Solubility in water: insoluble
Formula Weight 121.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Acetylpyridine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a
EINECSNumber 214-350-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.