Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Aminocaproic acid, 99+%

Catalog No. AC103300100
Click to view available options
Quantity:
100 g
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum

CAS: 60-32-2 | C6H13NO2 | 131.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-32-2
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.17
InChI Key SLXKOJJOQWFEFD-UHFFFAOYSA-N
Synonym 6-aminocaproic acid, aminocaproic acid, amicar, epsikapron, capramol, caprocid, epsamon, eaca, acepramin, caprolisin
PubChem CID 564
ChEBI CHEBI:16586
IUPAC Name 6-aminohexanoic acid
SMILES C(CCC(=O)O)CCN

Specifications

Melting Point 204.0°C to 209.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic drum
Linear Formula H2N(CH2)5CO2H
Quantity 10 kg
Beilstein 04,434
Merck Index 15,427
Solubility Information (13g/100ml H2O) Clear colorless solution
Formula Weight 131.17
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material 6-Aminocaproic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 200-469-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.